Learn More
Gears
Catalog No. S93923
Change view
Click to view available options
Quantity:
300 Gears
Catalog No. Quantity
S93923 300 Gears
1 options
Catalog No. S93923 Supplier Customer Channels Group (Ccg) Supplier No. EPD50001
Teach students all there is to know about gears.
  • Gear sets offer 40-, 30- and 20-tooth gears
  • With 3/16 bore to accommodate the most popular dowel size
  • Plus a 12-tooth gear with a push-fit to 2mm motor shafts
  • Three inserts are provided on each sprue, allowing the bore to reduce to 3mm
  • Gears are beveled for 90° meshing
  • 40-tooth gear has an additional hole to create an eccentric movement, or to use with a crank handle
Order Info
Available in 152 and 300 piece mixed color sets.
Specifications
For Use With (Application) Teach students all there is to know about gears
Includes 40, 30 and 20-tooth gears with 0.187 in. bore, 12-tooth gear with a push-fit to 2mm motor shafts, 300 gears in mixed colors set
Quantity 300 Gears
Product Type Gears Kit