missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ GDE Enzymatic Digester

Catalog No. 07222017
Change view
Click to view available options
Voltage:
115 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
07222017 115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07222017 Supplier Velp Scientifica™ Supplier No. SA30410209

missing translation for 'validMainframeMsg'

Enzymatic digester for dietary fiber analysis by enzymes heating.

  • Analytical procedure for the determination of dietary fiber involves a series of digestions of the sample by thermostable enzymes using the enzymatic digester GDE, which operates in accordance with the official AOAC method for the determination of total dietary fiber
  • Multi-position magnetic stirrer of GDE ensures homogeneity, thanks to a continuous and constant stirring of the sample while avoiding overheating that could compromise the success of enzymatic digestions
  • Transparent polycarbonate tank ensures visibility and enhanced resistance up to 105°C
  • Use of deionized water eliminates any possible creation of limestone
  • Outstanding temperature regulation is possible with GDE: through an intuitive digital display, it is possible to set the desired temperature, with high accuracy of only ±0.2°C

Specifications

Product Type Digestor
Display Digital
Voltage 115 V