Learn More
Thermo Scientific™ GC Tool Kits for Thermo Scientific Instruments
Catalog No. 60180784
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific and Agilent GC Instruments
Catalog No. For Use With (Equipment)
60180784 Thermo Scientific and Agilent GC Instruments
1 options
Catalog No. 60180784 Supplier Thermo Scientific™ Supplier No. 60180784
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Maintain the performance of your GC instrument with Thermo Scientific GC Tool Kits.
Maintain the performance of your GC system with Thermo Scientific™ GC Tool Kits. Wrench sets, a flashlight, brushes, a minidrill set, and other tools are included. Kits are available for Thermo Scientific GC and Agilent™ GC instruments.
Features
  • Replacements included perform like original instrument parts
  • Parts list included for easy reordering
Includes:
Capillary instrument gauge, liner cap removal tool, open-end wrenches in 6 x 7mm, 8 x 10mm and 16 x 17mm sizes.
Specifications
Type Capillary Tool Kit
Description GC Tool Kit for Thermo Scientific Instruments
For Use With (Application) GC
For Use With (Equipment) Thermo Scientific and Agilent GC Instruments
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.