missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ GC Stationary Phase, SP™-2100

Catalog No. 111002522
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002522 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002522 Supplier MilliporeSigma™ Supelco™ Supplier No. 21284U

A methyl silicone fluid suitable as stationary phase

  • Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase

Specifications

Max. Temperature 350°C
Stationary Phase SP™-2100
Packaging Bottle
Quantity 10 g
For Use With (Application) Suitable for G1
Physical Form Fluid

SP is a registered trademark of Sigma-Aldrich Co. LLC