missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ GC Stationary Phase, Dinonyl Phthalate

Catalog No. 111003010
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111003010 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111003010 Supplier MilliporeSigma™ Supelco™ Supplier No. 21052U

A general-purpose plasticizer for vinyl resins with low volatility and good electrical properties

  • Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase

Specifications

Max. Temperature 150°C
Stationary Phase Dinonyl Phthalate
Packaging Bottle
Quantity 50 g
Description Acetone: Soluble