missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ GC Stationary Phase, Carbowax 20M

Catalog No. 111002989
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002989 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002989 Supplier MilliporeSigma™ Supelco™ Supplier No. 21032

A polar, liquid phase suitable as a stationary phase for gas chromatography

  • This structure of the polymer makes it suitable for being used as a solvent for polar molecules such as cyclodextrin derivatives
  • It exhibits good selectivity toward alcohols, diols and aromatic isomeric molecules
  • Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase

Specifications

Max. Temperature 225°C
Stationary Phase Carbowax 20 M
Packaging Bottle
Quantity 50 g
Description Chloroform: Soluble
For Use With (Application) Suitable for G16
Physical Form Liquid