missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrodin 98.0+%, TCI America™
$267.70 - $801.21
Chemical Identifiers
| CAS | 62499-27-8 |
|---|---|
| Molecular Formula | C13H18O7 |
| Molecular Weight (g/mol) | 286.28 |
| MDL Number | MFCD00272169 |
| InChI Key | PUQSUZTXKPLAPR-UJPOAAIJSA-N |
| Synonym | 4-(Hydroxymethyl)phenyl beta-D-Glucopyranoside |
| PubChem CID | 115067 |
| ChEBI | CHEBI:80828 |
| IUPAC Name | (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[4-(hydroxymethyl)phenoxy]oxane-3,4,5-triol |
| SMILES | C1=CC(=CC=C1CO)OC2C(C(C(C(O2)CO)O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
G0468200MG
|
TCI America
G0468200MG |
200 mg |
Each for $267.70
|
|
|||||
|
G04681G
|
TCI America
G04681G |
1 g |
Each for $801.21
|
|
|||||
Chemical Identifiers
| 62499-27-8 | |
| 286.28 | |
| PUQSUZTXKPLAPR-UJPOAAIJSA-N | |
| 115067 | |
| (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[4-(hydroxymethyl)phenoxy]oxane-3,4,5-triol |
| C13H18O7 | |
| MFCD00272169 | |
| 4-(Hydroxymethyl)phenyl beta-D-Glucopyranoside | |
| CHEBI:80828 | |
| C1=CC(=CC=C1CO)OC2C(C(C(C(O2)CO)O)O)O |
Specifications
| 62499-27-8 | |
| White | |
| MFCD00272169 | |
| 4-(Hydroxymethyl)phenyl beta-D-Glucopyranoside | |
| C1=CC(=CC=C1CO)OC2C(C(C(C(O2)CO)O)O)O | |
| 286.28 | |
| CHEBI:80828 | |
| ≥98.0% (HPLC) | |
| Gastrodin |
| 155°C | |
| C13H18O7 | |
| 200 mg | |
| PUQSUZTXKPLAPR-UJPOAAIJSA-N | |
| (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[4-(hydroxymethyl)phenoxy]oxane-3,4,5-triol | |
| 115067 | |
| 286.28 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : LZ5776885
TSCA : No
Recommended Storage : Refrigerator