missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrodin 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 155°C |
| Color | White |
| Quantity | 200 mg |
| Formula Weight | 286.28 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Gastrodin |
Safety and Handling
| RTECSNumber | LZ5776885 |
| TSCA | No |
| Recommended Storage | Refrigerator |