missing translation for 'onlineSavingsMsg'
Learn More

BD GasPak™ Supplies

Catalog No. p-4162720
Change view
Click to view available options
Type:
CO2 Indicator Strip
Dry Anaerobic Indicator Strip
Quantity:
100 Pk.
50 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
B4371055 CO2 Indicator Strip 50 Pk.
B71051 Dry Anaerobic Indicator Strip 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4371055 Supplier BD Supplier No. 271055
Only null left

Used with the GasPak™ EZ Gas Generating Systems. BD™ GasPak™ Supplies are used with these complete, integrated systems for rapid, waterless and catalyst-free gas generation. Can be used to support an anaerobic, microaerophilic or CO2 enriched environment.

GasPak™ EZ Gas Generating Systems are complete, integrated systems that rapidly produce atmospheres suitable to support the primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets inside multi-use incubation containers.

Accessories include:

  • CO2 Indicator Strips
  • Dry Anaerobic Indicator Strips
  • Tube Holder
  • Catalyst Replacement Charges
  • Catalyst Reaction Chamber

Specifications

Type CO2 Indicator Strip
Content And Storage 15°C to 30°C
Description Indicator signifies that the carbon dioxide concentration in the atmosphere is 3% or greater.
Format Strip
For Use With (Application) Produce atmospheres suitable to support the primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria
For Use With (Equipment) BD GasPak™
Quantity 50 Pk.