missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Gas-tight Option Model

Includes additional measures for the greatest possible impermeability to gas; not in conjunction with access port or interior lighting option.

$4,118.58 - $4,370.68

Specifications

Description Gas-tight Option Model
Material Steel
Weight (English) 2.20 lb.
Weight (Metric) 1 kg
View More Specs

Products 3
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
01672907
View Documents Promotion Details
BINDER™
80122310
Units M 260
Each for $4,118.58
Only null left
 
01672908
View Documents Promotion Details
BINDER™
80122309
Units M 115
Each for $4,118.58
Only null left
 
01672909
View Documents Promotion Details
BINDER™
80122308
Units M 56
Each for $4,370.68
Only null left
 
Description

Description

  • Gas-tight option model
  • For use with various BINDER™ Unit M lines
Specifications

Specifications

Gas-tight Option Model
Steel
2.20 lb.
1 kg
Videos
SDS
Documents

Documents