Gas Dispersion Tube, Fine Frit. Hollow fritted cylinder at the end of an 8mm O.D. tube. Cylinder is approximately 12mm O.D. x 20mm long. Overall length is approximately 10 inches.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Gas Dispersion Tube, Fine Frit. Hollow fritted cylinder at the end of an 8mm O.D. tube. Cylinder is approximately 12mm O.D. x 20mm long. Overall length is approximately 10 inches.