missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Gas Chromatography Tubing

Catalog No. 03100082
Change view
Click to view available options
Material:
Copper
Stainless Steel
Diameter (English) Inner:
0.065 in.
0.085 in.
Diameter (English) Outer:
1/8 in.
Description:
25 ft. Length
50 ft. Length
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Diameter (English) Inner Diameter (English) Outer Description
03100082 Copper 0.065 in. 1/8 in. 50 ft. Length
03100088 Stainless Steel 0.085 in. 1/8 in. 25 ft. Length
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03100082 Supplier Thermo Scientific™ Supplier No. 60181632

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Find the copper and stainless steel tubing in a variety of sizes for plumbing your gas chromatography (GC) system.

Find copper and stainless steel Thermo Scientific™ Gas Chromatography Tubing in a variety of sizes for plumbing your gas chromatography (GC) system.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications

Type Gas Chromatography Tubing
Material Copper
Description 50 ft. Length
Diameter (English) Inner 0.065 in.
Diameter (English) Outer 1/8 in.
For Use With (Equipment) Gas Chromatography Systems

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.