missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Garage Physics Spiral of Theodorus Kit

Catalog No. S24328
Change view
Click to view available options
Spiral of Theodorus Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S24328 Spiral of Theodorus Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24328 Supplier Eisco™ Supplier No. GP00006

Allows students to experiment with square roots and irrational numbers in physical form.

  • Includes 18 wooden traingles to build Spiral of Theodorus, labels, pen, instruction guide
  • Hands-on activities aid in conceptualization of abstract mathematical ideas
  • Elementary school children use triangles to explore Pythagorean theorem and square roots
  • Middle school students use triangles to test equivalencies with square roots and explore abstract concepts like sequence of geometric shapes based on Fibonacci numbers
  • High school students use kit to explore proof of irrationality for non-square integer roots

Specifications

Product Type Kit, Spiral of Theodorus
Grade K to 12
For Use With (Application) Explore triangular relations
Includes 18 wooden triangles, labels, pen, and instruction guide
Grade Level Elementary, Intermediate - Middle School, High School