missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Garage Physics Murlin Trebuchet Kit

Catalog No. S88359
Change view
Click to view available options
Grade:
1 to 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S88359 1 to 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S88359 Supplier Eisco™ Supplier No. GP00015

Garage Physics Murlin Trebuchet Kit explores how the use of a simple machine (pulley) efficiently redirects gravitational potential energy to throw objects farther than a traditional trebuchet

By redirecting the pulling force of the counterweight with a pulley, the Garage Physics MURLIN Trebuchet kit is able to transfer much more energy to the projectile than the standard trebuchet.

  • Assembled kit throws the included ping pong ball 30 feet or more.
  • Manufactured with high quality, precision-cut Birch veneer pure bond plywood
  • Includes the trebuchet, ping pong ball, and 1 kg counterweight
  • A small Phillip's screwdriver is required for assembly
  • Kit comes with a step-by-step educational guide

Warranty

1 year limited manufacturer warranty

Specifications

Product Type Physics tool
Grade 1 to 8
Material Wood
For Use With (Application) Classroom demonstration
Includes Trebuchet, Ping pong ball, Counterweight, Manual
Quantity 1
Color Brown