missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Garage Physics Magic Blocks Kit

Catalog No. S24327
Change view
Click to view available options
Magic blocks kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S24327 Magic blocks kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24327 Supplier Eisco™ Supplier No. GP00005
Only null left

Hands-on activities kit aid in the conceptualization of abstract mathematical ideas.

  • Nine rectangular wood blocks of steadily increasing height
  • For all ages of children, from youngsters to hight school
  • Young children use kit to explore natural numbers
  • Elementary school children use to investigate addition, subtraction, multiplication, units of measure
  • Middle school students explore magic square in depth and arthimetic sequences
  • High school students explore counting in binary up to 15 and in ternary up to 26

Specifications

Product Type Kit, Magic Blocks
Grade K to 12
For Use With (Application) Explore basic mathematical operators and concepts
Includes Nine rectangular blocks of steadily increasing height, pen, labels, and instructions
Grade Level Elementary, Intermediate - Middle School, High School