Learn More
Eisco™ Garage Physics Large Trebuchet DIY Kit
Catalog No. S43526
Change view
Click to view available options
Large Trebuchet DIY Kit
Catalog No.
S43526 Large Trebuchet DIY Kit
1 options
Catalog No. S43526 Supplier Eisco™ Supplier No. GP00009
Kit engages the young learner to discover new ideas about the physical world through assembly and play. EISCO Garage Physics Large Trebuchet DIY Kit kits contain pre-cut wooden pieces that resemble woodworking projects.
  • Medieval weapons, engineering, and physics - not a bad way to spend a Saturday afternoon constructing this trebuchet which will throw the included ping pong ball more than 30 feet and heavier objects farther.
  • The main material is high-quality precision-cut Birch vernier pure bond plywood.
  • All necessary parts are included and holes are pre-drilled.
  • The kit is specially designed to be safe and robust. Dry firings and heavy projectiles are no problems for this kit.
  • All that is needed for assembly is time, a screwdriver, a hammer, and fun. No glue, mess or fuss, just assembly, and experimentation
The trebuchet is one of the most elegant of all medieval weapons ever invented. With a simple counterweight, beam arm, and sling, heavier projectiles can be launched much further than is possible with other siege engines. Think of it as a machine which transforms gravitational potential energy into kinetic energy. A lot of kinetic energy! Arcing across your yard! We include a ping pong ball, but have experimented with marshmallows, jelly beans, bouncy balls and other safe objects. Two release pin angles allow exploration of how the release angle changes the trajectory. The included guide provides clear assembly instructions with pictures. It also contains pedagogy for exploring concepts of energy transfer using the scientific method.
Specifications
Product Type Trebuchet DIY Kit
Description 21 in. beam arm
Material Birch Veneer Pure Bond Plywood
For Use With (Application) Medieval weapons, engineering, and physics studies
Weight (English) 4.7 lb.
Electrical Requirements None