missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Garage Physics Catapult Kit
Description
- Designed for elementary through high school children
- Contains catapult base, sides, lever arm, rubber bands, screws, dowels, hook, and ping-pong ball
- DIY kit includes adjustable parts to experiment with different initial conditions
- Instructions detail learning exercises and routes of further exploration
Specifications
Specifications
| Product Type | Kit, Catapult |
| Grade | K to 12 |
| For Use With (Application) | Explore force, acceleration, Hooke's law, and parabolic trajectories |
| Includes | Catapult base, sides, lever arm, rubber bands, screws, dowels, hook, and ping-pong ball |
| Grade Level | Elementary, Intermediate - Middle School, High School |