missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Garage Physics Ball Bearing Roller Coaster Kit

Catalog No. S88361
Change view
Click to view available options
Grade:
1 to 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S88361 1 to 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S88361 Supplier Eisco™ Supplier No. GP00017

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Garage Physics Ball Bearing Roller Coaster Kit for students and educators studying gravitational potential energy, translational and rotational kinetic energy

  • Allows students to watch potential energy convert to kinetic energy
  • Easy to assemble
  • Includes educational guide
  • Kit Includes:
    • 20-foot long (5/8 in. diameter) FDA approved vinyl tube
    • 9 removable wire hooks
    • Twist ties and clipper
    • 4 steel ball bearings (3/8 in. diameter)
    • 3 rubbing alcohol pads
  • Stick included hooks to the wall or other vertical surface, or use chairs and tables to support the track
  • Create loop de loops, corkscrews and dives
  • Required but not included:
    • Scratch paper (US Letter or A4 size is sufficient)
    • Pair of sissors
    • Tape
    • 2 tin cans per class (any size)

Warranty

1 year limited manufacturer warranty

Specifications

Product Type Physics
Grade 1 to 8
Material Rubber, Metal
For Use With (Application) Classroom demonstration
Includes 20 ft tubing, (4) ball bearings, twisty ties, (3)rubbing alcohol pads, (9)hooks, manual
Quantity 1
Color Clear