Recombinant Protein;Human GAPDH / G3PDH Protein;1MG;Human GAPDH, His Tag (GAH-H5145) is expressed from E.coli cells. It contains AA Gly 2 - Glu 335 (Accession # AAH01601).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 36.9 kDa. The protein migrates as 37 kDa under reducing (R) condition (SDS-PAGE).
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Human GAPDH / G3PDH Protein;1MG;Human GAPDH, His Tag (GAH-H5145) is expressed from E.coli cells. It contains AA Gly 2 - Glu 335 (Accession # AAH01601).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 36.9 kDa. The protein migrates as 37 kDa under reducing (R) condition (SDS-PAGE).