missing translation for 'onlineSavingsMsg'
Learn More

Ansell GAMMEX™ Cut-Resistant Glove Liners

Catalog No. 17103767
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17103767 X-Small
17103768 Small
17103769 Medium
17103770 Large
17103771 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17103767 Supplier Ansell Supplier No. 5789910

Non-latex, Diamond yarn cut-resistant glove liners provide adequate cut resistance and protection during rigorous procedures

  • ANSI A2 Cut Rating: Cut protection during rigorous, high-risk procedures
  • Lightweight Dyneema Diamond Yarn: Uncompromising comfort and dexterity
  • Allergy Optimized Formulation: Non-latex and 100% chemical accelerator-free
  • Adequate cut-resistance
  • Sterile

Specifications

ANSI Abrasion Rating ANSI 2
ASTM F739 No
Chlorinated No
Construction Knit
Cuff Material Uncoated
Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Latex Free Yes
Material Nylon
Product Type Cut-resistant Glove Liners
Size X-Small
Sterility Sterile
EN 421 No
FDA 510 (k) No
For Use With (Application) Offer light to moderate level cut protection from laceration-type and scalpel injuries that may occur during rigorous procedures
For Use With (Equipment) In between two surgical gloves
Gender Unisex
Glove Cuff Color Orange
Glove Cuff Style Knit
Lining Unlined
Thumb Style Straight
Show More Show Less