missing translation for 'onlineSavingsMsg'
Learn More

gamma-Valerolactone 98.0+%, TCI America™

Catalog No. V000725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V000725G 25 g
V0007500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V000725G Supplier TCI America Supplier No. V000725G

Chemical Identifiers

CAS 108-29-2
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00005400
InChI Key GAEKPEKOJKCEMS-UHFFFAOYSA-N
Synonym gamma-valerolactone, 4-valerolactone, 4-pentanolide, 5-methyldihydrofuran-2 3h-one, 4-hydroxypentanoic acid lactone, gamma-pentalactone, 4-hydroxyvaleric acid lactone, 2 3h-furanone, dihydro-5-methyl, 4-methyl-gamma-butyrolactone, pentanolide-1,4
PubChem CID 7921
ChEBI CHEBI:48569
IUPAC Name 5-methyloxolan-2-one
SMILES CC1CCC(=O)O1

Specifications

Melting Point -31°C
Color Yellow
Boiling Point 207°C
Quantity 25 g
Formula Weight 100.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material gamma-Valerolactone
EINECSNumber (9)-0137
RTECSNumber LU3580000
TSCA Yes