missing translation for 'onlineSavingsMsg'
Learn More

gamma-Terpinene 95.0+%, TCI America™

Catalog No. M0318500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0318500ML 500 mL
M031825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0318500ML Supplier TCI America Supplier No. M0318500ML

Chemical Identifiers

CAS 99-85-4
Molecular Formula C10H16
Molecular Weight (g/mol) 136.238
MDL Number MFCD00001537
InChI Key YKFLAYDHMOASIY-UHFFFAOYSA-N
Synonym gamma-terpinene, p-mentha-1,4-diene, crithmene, moslene, gamma-terpinen, 1,4-p-menthadiene, 4-isopropyl-1-methyl-1,4-cyclohexadiene, 1,4-cyclohexadiene, 1-methyl-4-1-methylethyl, terpinene, alpha, .gamma.-terpinen
PubChem CID 7461
ChEBI CHEBI:10577
IUPAC Name 1-methyl-4-propan-2-ylcyclohexa-1,4-diene
SMILES CC1=CCC(=CC1)C(C)C

Specifications

Color Yellow
Boiling Point 183°C
Quantity 500 mL
UN Number 2319
Formula Weight 136.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material gamma-Terpinene
EINECSNumber (3)-2244
RTECSNumber OS8070000
TSCA Yes