missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma-Phenyl-gamma-butyrolactone 97.0+%, TCI America™
Supplier: TCI America P132125G
Specifications
| gamma-Phenyl-gamma-butyrolactone | |
| 36°C | |
| 306°C | |
| MFCD00005398 | |
| gamma-phenyl-gamma-butyrolactone, 4-phenylbutanolide, 4-phenyl-4-butanolide, 4-phenylbutyrolactone, 2 3h-furanone, dihydro-5-phenyl, 5-phenyldihydrofuran-2 3h-one, 4-phenyl-4-hydroxybutanoic acid lactone, 4,5-dihydro-5-phenyl-2 3h-furanone, .gamma.-phenyl-.gamma.-butyrolactone, .gamma.-phenylbutyrolactone | |
| O=C1CCC(O1)C1=CC=CC=C1 | |
| 162.19 | |
| 162.19 | |
| Crystalline Powder |
| 1008-76-0 | |
| White | |
| C10H10O2 | |
| 25 g | |
| AEUULUMEYIPECD-UHFFFAOYNA-N | |
| 5-phenyloxolan-2-one | |
| 13884 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 1008-76-0 | |
| 162.19 | |
| AEUULUMEYIPECD-UHFFFAOYNA-N | |
| 13884 | |
| O=C1CCC(O1)C1=CC=CC=C1 |
| C10H10O2 | |
| MFCD00005398 | |
| gamma-phenyl-gamma-butyrolactone, 4-phenylbutanolide, 4-phenyl-4-butanolide, 4-phenylbutyrolactone, 2 3h-furanone, dihydro-5-phenyl, 5-phenyldihydrofuran-2 3h-one, 4-phenyl-4-hydroxybutanoic acid lactone, 4,5-dihydro-5-phenyl-2 3h-furanone, .gamma.-phenyl-.gamma.-butyrolactone, .gamma.-phenylbutyrolactone | |
| 5-phenyloxolan-2-one |
Safety and Handling
RTECSNumber : LU3690000
TSCA : No