missing translation for 'onlineSavingsMsg'
Learn More

gamma-Phenyl-gamma-butyrolactone 97.0+%, TCI America™

Catalog No. P13215G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13215G 5 g
P132125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13215G Supplier TCI America Supplier No. P13215G
Only null left

Chemical Identifiers

CAS 1008-76-0
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00005398
InChI Key AEUULUMEYIPECD-UHFFFAOYNA-N
Synonym gamma-phenyl-gamma-butyrolactone, 4-phenylbutanolide, 4-phenyl-4-butanolide, 4-phenylbutyrolactone, 2 3h-furanone, dihydro-5-phenyl, 5-phenyldihydrofuran-2 3h-one, 4-phenyl-4-hydroxybutanoic acid lactone, 4,5-dihydro-5-phenyl-2 3h-furanone, .gamma.-phenyl-.gamma.-butyrolactone, .gamma.-phenylbutyrolactone
PubChem CID 13884
IUPAC Name 5-phenyloxolan-2-one
SMILES O=C1CCC(O1)C1=CC=CC=C1

Specifications

Melting Point 36°C
Color White
Boiling Point 306°C
Quantity 5 g
Formula Weight 162.19
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material gamma-Phenyl-gamma-butyrolactone
RTECSNumber LU3690000
TSCA No