missing translation for 'onlineSavingsMsg'
Learn More

gamma-Octanolactone, 98%

CAS: 104-50-7 | C8H14O2 | 142.198 g/mol

Supplier:  Thermo Scientific Chemicals A1814222

 View more versions of this product

Catalog No. AAA1814222

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

gamma-Octanolactone
0.981
126°C (259°F)
1.444
100 g
IPBFYZQJXZJBFQ-UHFFFAOYSA-N
5-butyloxolan-2-one
7704
98%
104-50-7
233°C to 235°C
C8H14O2
MFCD00005402
gamma-octalactone, 4-octanolide, 5-butyldihydrofuran-2 3h-one, octanolide-1,4, 2 3h-furanone, 5-butyldihydro, octan-4-olide, gamma-octanolactone, 8-oxo-5-octanolide, gamma-octanoic lactone, 5-butyltetrahydro-2-furanone
CCCCC1CCC(=O)O1
142.198
142.2

Chemical Identifiers

104-50-7
142.198
IPBFYZQJXZJBFQ-UHFFFAOYSA-N
7704
CCCCC1CCC(=O)O1
C8H14O2
MFCD00005402
gamma-octalactone, 4-octanolide, 5-butyldihydrofuran-2 3h-one, octanolide-1,4, 2 3h-furanone, 5-butyldihydro, octan-4-olide, gamma-octanolactone, 8-oxo-5-octanolide, gamma-octanoic lactone, 5-butyltetrahydro-2-furanone
5-butyloxolan-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H320-H335

EINECSNumber : 203-208-1

RTECSNumber : LU3562000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only