missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma-Octanoic lactone, 98%
CAS: 104-50-7 | C8H14O2 | 142.2 g/mol
$68.22 - $68.22
Chemical Identifiers
| CAS | 104-50-7 |
|---|---|
| Molecular Formula | C8H14O2 |
| Molecular Weight (g/mol) | 142.2 |
| MDL Number | MFCD00005402 |
| InChI Key | IPBFYZQJXZJBFQ-UHFFFAOYSA-N |
| Synonym | gamma-octalactone, 4-octanolide, 5-butyldihydrofuran-2 3h-one, octanolide-1,4, 2 3h-furanone, 5-butyldihydro, octan-4-olide, gamma-octanolactone, 8-oxo-5-octanolide, gamma-octanoic lactone, 5-butyltetrahydro-2-furanone |
| PubChem CID | 7704 |
| IUPAC Name | 5-butyloxolan-2-one |
| SMILES | CCCCC1CCC(=O)O1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC129400250
|
Thermo Scientific Chemicals
129400250 |
25 g | Glass bottle |
Each for $68.22
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 104-50-7 | |
| 142.2 | |
| IPBFYZQJXZJBFQ-UHFFFAOYSA-N | |
| 7704 | |
| CCCCC1CCC(=O)O1 |
| C8H14O2 | |
| MFCD00005402 | |
| gamma-octalactone, 4-octanolide, 5-butyldihydrofuran-2 3h-one, octanolide-1,4, 2 3h-furanone, 5-butyldihydro, octan-4-olide, gamma-octanolactone, 8-oxo-5-octanolide, gamma-octanoic lactone, 5-butyltetrahydro-2-furanone | |
| 5-butyloxolan-2-one |
Specifications
| 104-50-7 | |
| 0.9800g/mL | |
| >110°C | |
| 97.5% min. (GC) | |
| C8H14O2 | |
| MFCD00005402 | |
| 0.98 | |
| IPBFYZQJXZJBFQ-UHFFFAOYSA-N | |
| 5-butyloxolan-2-one | |
| 7704 | |
| 98% | |
| γ-Octanoic lactone, 98% |
| Colorless | |
| 234.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4430 to 1.4450 | |
| 25 g | |
| gamma-octalactone, 4-octanolide, 5-butyldihydrofuran-2 3h-one, octanolide-1,4, 2 3h-furanone, 5-butyldihydro, octan-4-olide, gamma-octanolactone, 8-oxo-5-octanolide, gamma-octanoic lactone, 5-butyltetrahydro-2-furanone | |
| CCCCC1CCC(=O)O1 | |
| 142.2 | |
| 142.2 | |
| Liquid |
Safety and Handling
EINECSNumber : 203-208-1
RTECSNumber : LU3562000
TSCA : TSCA
RUO – Research Use Only