missing translation for 'onlineSavingsMsg'
Learn More

gamma-Nonanolactone 98.0+%, TCI America™

Catalog No. n028525ml
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N028525ML 25 mL
N0285500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N028525ML Supplier TCI America Supplier No. N028525ML
Only null left

Chemical Identifiers

CAS 104-61-0
Molecular Formula C9H16O2
Molecular Weight (g/mol) 156.23
MDL Number MFCD00005403
InChI Key OALYTRUKMRCXNH-QMMMGPOBSA-N
Synonym gamma-Nonalactone, gamma-Pelargonolactone
PubChem CID 7710
IUPAC Name (5S)-5-pentyloxolan-2-one
SMILES CCCCC[C@H]1CCC(=O)O1

Specifications

Color Colorless
Boiling Point 243°C
Quantity 25 mL
Formula Weight 156.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material gamma-Nonanolactone
EINECSNumber (9)-0137
RTECSNumber LU3675000
TSCA Yes