missing translation for 'onlineSavingsMsg'
Learn More

gamma-Methylene-gamma-butyrolactone 98.0+%, TCI America™

Catalog No. M13851G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M13851G Supplier TCI America Supplier No. M13851G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10008-73-8
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.10
MDL Number MFCD00042762
InChI Key SIFBVNDLLGPEKT-UHFFFAOYSA-N
Synonym gamma-methylene-gamma-butyrolactone, dihydro-5-methylenefuran-2 3h-one, unii-b6fm50xsnz, alpha'-angelica lactone, 5-methylenedihydrofuran-2 3h-one, b6fm50xsnz, 4-hydroxy-4-pentenoic acid gamma-lactone, 5-methylenedihydro-2 3h-furanone, 2 3h-furanone, dihydro-5-methylene, chembl93112
PubChem CID 66201
ChEBI CHEBI:36439
IUPAC Name 5-methylideneoxolan-2-one
SMILES C=C1CCC(=O)O1

Specifications

Color Colorless
Quantity 1 g
Physical Form Clear Liquid at 20°C
Formula Weight 98.10
Percent Purity ≥98.0% (GC)
Chemical Name or Material gamma-Methylene-gamma-butyrolactone