missing translation for 'onlineSavingsMsg'
Learn More

Keystone Cleanroom Products Gamma Irradiated Trash Bag

Pre-sterilized trash bags for pharmaceutical cleanrooms. Simplify the removal of waste from your aseptic cleanroom without introducing external sources of microbial contamination.

Supplier:  Keystone Cleanroom Products S3655CP5

 View more versions of this product

Catalog No. 19990236


Only null left

Description

Description

  • Packaged in an ISO6 cleanroom
  • 2 mil clear polyethylene
  • Each package includes a lot number for traceability
  • Certificates of Conformance and Irradiation sent with shipment
  • Packaged in Tyvek™/Mylar Chevron Peel Pouches
Specifications

Chemical Identifiers

Not Autoclavable
None
Single-use
Trash Bin
Sterile
2 mil
36 in.
15 gal.
139.7 cm
0.05 mm
COC, Certificate of Irradiation
Clear
Cleaning
LDPE
Gamma Irradiated
55 in.
Cleanroom Bags
56.9 L
91.4 cm

Specifications

Not Autoclavable
None
Single-use
Trash Bin
Sterile
Gamma Irradiated
55 x 36 in.
36 in.
15 gal.
139.7 cm
0.05 mm
COC, Certificate of Irradiation
Clear
Cleaning
LDPE
5 Pk.
2 mil
55 in.
Cleanroom Bags
56.9 L
91.4 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions