Learn More
Keystone Cleanroom Products Gamma-Irradiated Retractable Ultra Fine Point Sharpie
Catalog No. 17010189
Change view
Click to view available options
Color:
Black
Blue
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17010189 Black
17010190 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17010189 Supplier Keystone Cleanroom Products Supplier No. S01RETMARKBLK
Ready-to-use gamma irradiated pens and markers eliminate the risks associated with surface sanitization procedures.
  • Gamma irradiated
  • Lot controlled
  • Ready to use
  • Packaged in easy to open Tyvek™ peel pouches
  • Minimize contamination risks
  • Simplifies conformance to SOP's
  • Eliminate the need for wiping down contaminated writing instruments
Specifications
Tip Style Ultrafine
Product Type Cleanroom Marker
Color Black
Autoclavable Not Autoclavable
Sterility Sterile
Material Plastic
Disposable Single-use
Certifications/Compliance COC, Certificate of Irradiation
Includes Retractable
No. per Case 250/Cs.
Packaging Type Peel Pouch
Show More Show Less