missing translation for 'onlineSavingsMsg'
Learn More

gamma-Decanolactone 96.0+%, TCI America™

Catalog No. H023325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H023325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H023325G Supplier TCI America Supplier No. H023325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 706-14-9
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00005404
InChI Key IFYYFLINQYPWGJ-UHFFFAOYNA-N
Synonym gamma-decalactone, 4-decanolide, gamma-decanolactone, 2 3h-furanone, 5-hexyldihydro, 5-hexyldihydrofuran-2 3h-one, decan-4-olide, 5-hexyldihydro-2 3h-furanone, decanolide-1,4, 2-decalactone, 4-hexyl-4-butanolide
PubChem CID 12813
IUPAC Name 5-hexyloxolan-2-one
SMILES CCCCCCC1CCC(=O)O1

Specifications

Color Colorless
Boiling Point 281°C
Quantity 25 g
Formula Weight 170.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material gamma-Decanolactone
EINECSNumber (9)-0137&(5)-1173
RTECSNumber LU4600000
TSCA Yes