missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma-Cyclodextrin 99.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 100 g |
| Formula Weight | 1297.13 |
| Percent Purity | ≥99.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | gamma-Cyclodextrin |
Safety and Handling
| EINECSNumber | (8)-0101 |
| RTECSNumber | GU2293080 |
| TSCA | Yes |