missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma-Cyclodextrin, Approx. 98%, Spectrum™ Chemical
C1623, 17465-86-0, C48H80O40
Supplier: Spectrum Chemical Mfg Cor C16231GM
Description
Spectrum™ Chemical gamma-Cyclodextrin , also known as Cyclodextrins, are used in the food, pharmaceutical, and chemical industries because they enhance the solubility and bioavailability of such compounds. This is of high interest for pharmaceutical as well as dietary supplement applications in which hydrophobic compounds shall be delivered. Alpha-, beta-, and gamma-cyclodextrin are all generally recognized as safe by the FDA. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Specifications
| 17465-86-0 | |
| C48H80O40 | |
| 1 g | |
| OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC9C(CO)OC(OC1C(O)C2O)C(O)C9O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O | |
| 1297.13 | |
| Reagent |
| 100% | |
| MFCD00009595,MFCD00149574 | |
| GDSRMADSINPKSL-UHFFFAOYNA-N | |
| 5,10,15,20,25,30,35,40-octakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,â¶.2â¸,¹¹.2¹³,¹â¶.2¹â¸,²¹.2²³,²â¶.2²â¸,³¹.2³³,³â¶]hexapentacontane-41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecol | |
| ∼98% | |
| Amber Glass Bottle |
Chemical Identifiers
| 17465-86-0 | |
| 1297.13 | |
| GDSRMADSINPKSL-UHFFFAOYNA-N | |
| OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC9C(CO)OC(OC1C(O)C2O)C(O)C9O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O |
| C48H80O40 | |
| MFCD00009595,MFCD00149574 | |
| 5,10,15,20,25,30,35,40-octakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,â¶.2â¸,¹¹.2¹³,¹â¶.2¹â¸,²¹.2²³,²â¶.2²â¸,³¹.2³³,³â¶]hexapentacontane-41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecol |