Learn More
Vikan™ Galvanized Steel Undercarriage
Description
- Galvanized steel
- Low undercarriage
- Features caster wheels that are impervious to meat fat and bearings equipped with grease fittings
- The zinc coating on the steel keeps the undercarriage free from rust, even in very moist environments
- A low undercarriage is a great solution when mobility is required
Specifications
Specifications
| Product Type | Undercarriage |
| Color | Steel |
| Material | Galvanized Steel |
| Quantity | 1 |
| Dimensions (L x W x H) | 42.5 x 21.6 x 9.4 in. |