missing translation for 'onlineSavingsMsg'
Learn More

(-)-Gallocatechin

Catalog No. AAJ60731MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60731MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60731MA Supplier Thermo Scientific Chemicals Supplier No. J60731MA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3371-27-5 | C15H14O7 | 306.27 g/mol

Polyphenolic compound from green tea. Possesses free-radical scavenging ability

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Polyphenolic compound from green tea
  • Possesses free-radical scavenging ability

Chemical Identifiers

CAS 3371-27-5
Molecular Formula C15H14O7
Molecular Weight (g/mol) 306.27
MDL Number MFCD01632616
InChI Key XMOCLSLCDHWDHP-DOMZBBRYSA-N
Synonym --gallocatechin, 2s,3r-2-3,4,5-trihydroxyphenyl chroman-3,5,7-triol, 2s,3r-gallocatechin, ent-gallocatechin, 2s,3r-flavan-3,3',4',5,5',7-hexol, 2s,3r-flavan-3,5,7,3',4',5'-hexol, 2s,3r-2-3,4,5-trihydroxyphenyl-3,4-dihydro-2h-chromene-3,5,7-triol, 2s,3r---2-3,4,5-trihydroxyphenyl-3,4-dihydro-2h-chromene-3,5,7-triol, gallocatechingc, l-gallocatechin
PubChem CID 9882981
ChEBI CHEBI:71225
IUPAC Name (2S,3R)-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)O

Specifications

Color White
Quantity 10 mg
Formula Weight 306.27
Physical Form Powder
Chemical Name or Material (-)-Gallocatechin
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only