missing translation for 'onlineSavingsMsg'
Learn More

Gallium pellets, 99.9999% (metals basis)

CAS: 7440-55-3 | Ga | 69.72 g/mol

$257.74 - $257.74

Chemical Identifiers

CAS 7440-55-3
Molecular Formula Ga
Molecular Weight (g/mol) 69.72
MDL Number MFCD00134045
InChI Key GYHNNYVSQQEPJS-UHFFFAOYSA-N
Synonym gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom
PubChem CID 5360835
ChEBI CHEBI:49631
IUPAC Name gallium
SMILES [Ga]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4432409
View Documents
Thermo Scientific Chemicals
04432409
10 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-55-3
69.72
GYHNNYVSQQEPJS-UHFFFAOYSA-N
5360835
gallium
Ga
MFCD00134045
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom
CHEBI:49631
[Ga]

Specifications

7440-55-3
99.9999%, metal basis
Ga
gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom
[Ga]
69.72
CHEBI:49631
10 g
Pellet
MFCD00134045
GYHNNYVSQQEPJS-UHFFFAOYSA-N
gallium
5360835
Gallium
Videos
Safety and Handling

Safety and Handling

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : DOT Class: 8; Packaging Group: III

EINECSNumber : 231-163-8

RTECSNumber : LW8600000

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only