Learn More
Gallium pellets, 6mm dia, 99.99999% (metals basis)
CAS: 7440-55-3 | Ga | 69.72 g/mol
$854.41 - $854.41
Chemical Identifiers
| CAS | 7440-55-3 |
|---|---|
| Molecular Formula | Ga |
| Molecular Weight (g/mol) | 69.72 |
| MDL Number | MFCD00134045 |
| InChI Key | GYHNNYVSQQEPJS-UHFFFAOYSA-N |
| Synonym | gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom |
| PubChem CID | 5360835 |
| ChEBI | CHEBI:49631 |
| IUPAC Name | gallium |
| SMILES | [Ga] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-55-3 | |
| 69.72 | |
| GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
| 5360835 | |
| gallium |
| Ga | |
| MFCD00134045 | |
| gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom | |
| CHEBI:49631 | |
| [Ga] |
Specifications
| Gray-Silver | |
| 50 g | |
| Pellet | |
| MFCD00134045 | |
| gallium, elemental, metal, unii-ch46oc8yv4, ch46oc8yv4, ingot, pellets, 6mm dia, galio, metal, packaged in poylethylene bottle, gallium, ion ga1+, atom | |
| [Ga] | |
| 69.72 | |
| CHEBI:49631 | |
| Gallium pellets, 6mm dia. |
| 7440-55-3 | |
| Max. weight 0.7g/pellet | |
| Ga | |
| UN2803 | |
| GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
| gallium | |
| 5360835 | |
| 99.99999% | |
| (metals basis) |
Safety and Handling
GHS H Statement
H290
May be corrosive to metals.
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: GALLIUM
EINECSNumber : 231-163-8
RTECSNumber : LW8600000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only