missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) trifluoromethanesulfonate, 99%

Catalog No. AC343930010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC343930010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC343930010 Supplier Thermo Scientific Chemicals Supplier No. 343930010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 74974-60-0 | C3F9GaO9S3 | 516.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 74974-60-0
Molecular Formula C3F9GaO9S3
Molecular Weight (g/mol) 516.91
MDL Number MFCD02093898
InChI Key BVLXNPRUOXPBII-UHFFFAOYSA-K
Synonym gallium iii trifluoromethanesulfonate, gallium tris trifluoromethanesulfonate, gallium trifluoromethanesulfonate, gallium triflate, gallium iii trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, gallium salt, galium iii triflate, gallium iii triflate, gallium 3+ ion tritriflate, gallium tris fluoranyl methanesulfonate
PubChem CID 2733940
IUPAC Name gallium;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Ga+3]

Specifications

Color White
Density 1.7000g/mL
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ex F)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.7
Solubility Information Solubility in water: soluble in water
Formula Weight 516.91
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Gallium(III) trifluoromethanesulfonate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only