missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) oxide, Puratronic™, 99.999% (metals basis)

CAS: 12024-21-4 | Ga2O3 | 187.44 g/mol

Supplier:  Thermo Scientific Chemicals 04252814

 View more versions of this product

Catalog No. AA4252814

Only null left
Explore available promotions

Description

Description

Gallium(III) oxide is used in vacuum deposition. It is useful for making semiconductor devices, gallium-alumina catalyst, gas sensors, luminescent phosphors and dielectric coatings of solar cells. Used as an evaporated material and sputtering target of 99.999% in dielectric films. It shows potential for developing deep-ultraviolet TCOs (Transparent Conductive Oxides) and transparent electrodes for ultraviolet optoelectronic devices. Recent studies report that gallium oxide can be a strong contender for power electronic devices for example in ultrahigh-voltage power switching applications. Films made of gallium oxide have gained commercial interest owing to their gas sensitive characteristics, and glasses made with gallium oxide are the preferred optical materials for use in advanced technologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gallium (III) oxide
12024-21-4
25 g
Ga2O3
14,4346
AJNVQOSZGJRYEI-UHFFFAOYSA-N
digallium(3+) trioxidandiide
187.44
Puratronic
1.92
1900°C
Powder
(metals basis)
MFCD00011020
Insoluble in water. Soluble in most of the acids.
[O--].[O--].[O--].[Ga+3].[Ga+3]
187.44
99.999%
Odorless
6.44 g/mL

Chemical Identifiers

12024-21-4
187.44
AJNVQOSZGJRYEI-UHFFFAOYSA-N
[O--].[O--].[O--].[Ga+3].[Ga+3]
Ga2O3
MFCD00011020
digallium(3+) trioxidandiide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 234-691-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only