missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) oxide, Puratronic™, 99.999% (metals basis)

Catalog No. AA4252806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4252806 5 g
AA4252814 25 g
AA4252822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4252806 Supplier Thermo Scientific Chemicals Supplier No. 04252806

CAS: 12024-21-4 | Ga2O3 | 187.44 g/mol

Gallium(III) oxide is used in vacuum deposition. It is useful for making semiconductor devices, gallium-alumina catalyst, gas sensors, luminescent phosphors and dielectric coatings of solar cells. Used as an evaporated material and sputtering target of 99.999% in dielectric films. It shows potential for developing deep-ultraviolet TCOs (Transparent Conductive Oxides) and transparent electrodes for ultraviolet optoelectronic devices. Recent studies report that gallium oxide can be a strong contender for power electronic devices for example in ultrahigh-voltage power switching applications. Films made of gallium oxide have gained commercial interest owing to their gas sensitive characteristics, and glasses made with gallium oxide are the preferred optical materials for use in advanced technologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12024-21-4
Molecular Formula Ga2O3
Molecular Weight (g/mol) 187.44
MDL Number MFCD00011020
InChI Key AJNVQOSZGJRYEI-UHFFFAOYSA-N
IUPAC Name digallium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Ga+3].[Ga+3]

Specifications

Melting Point 1900°C
Physical Form Powder
Quantity 5 g
Merck Index 14,4346
Solubility Information Insoluble in water. Soluble in most of the acids.
Formula Weight 187.44
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Refractive Index 1.92
Density 6.44 g/mL
Assay (metals basis)
Chemical Name or Material Gallium (III) oxide
Show More Show Less
EINECSNumber 234-691-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only