missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) oxide, Puratronic™, 99.999% (metals basis)

Used in vacuum deposition. | CAS: 12024-21-4 | Ga2O3 | 187.44 g/mol

Supplier:  Thermo Scientific Chemicals 01115122

 View more versions of this product

Catalog No. AA1115122


Only null left
Explore available promotions

Description

Description

Gallium(III) oxide is used in vacuum deposition. It is useful for making semiconductor devices, gallium-alumina catalyst, gas sensors, luminescent phosphors and dielectric coatings of solar cells. It shows potential for developing deep-ultraviolet TCOs (Transparent Conductive Oxides) and transparent electrodes for ultraviolet optoelectronic devices. Recent studies report that gallium oxide can be a strong contender for power electronic devices for example, in ultrahigh-voltage power switching applications. Films made of gallium oxide have gained commercial interest owing to their gas sensitive characteristics, and glasses made with gallium oxide are the preferred optical materials for use in advanced technologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gallium(III) oxide
1900°C
Plastic bottle
1.92
100 g
Soluble in acids. Insoluble in water
[O--].[O--].[O--].[Ga+3].[Ga+3]
187.44
99.999%
12024-21-4
6.44
Ga2O3
MFCD00011020
6.44
AJNVQOSZGJRYEI-UHFFFAOYSA-N
digallium(3+) trioxidandiide
187.44
Powder

Chemical Identifiers

12024-21-4
187.44
AJNVQOSZGJRYEI-UHFFFAOYSA-N
[O--].[O--].[O--].[Ga+3].[Ga+3]
Ga2O3
MFCD00011020
digallium(3+) trioxidandiide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 234-691-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only