missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) oxide, 99.995% (metals basis)

Catalog No. p-7050255
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3631806 5 g
AA3631814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3631806 Supplier Thermo Scientific Chemicals Supplier No. 03631806
Only null left

For vacuum deposition | CAS: 12024-21-4 | Ga2O3 | 187.44 g/mol

For vacuum deposition. In the production of catalysts. Used as part of the manufacturing of semiconductor devices. Has been shown to demonstrate catalytic properties and has also been used as an insulating barrier in tight junctions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12024-21-4
Molecular Formula Ga2O3
Molecular Weight (g/mol) 187.44
MDL Number MFCD00011020
InChI Key AJNVQOSZGJRYEI-UHFFFAOYSA-N
IUPAC Name digallium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Ga+3].[Ga+3]

Specifications

Melting Point 1900°C
Physical Form Sintered Lump
Quantity 5 g
Merck Index 14,4346
Solubility Information Insoluble in water. Soluble in most acids.
Formula Weight 187.44
Percent Purity 99.995%
Odor Odorless
Refractive Index 1.92
Density 6.44 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) oxide
Show More Show Less
EINECSNumber 234-691-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only