missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallium(III) oxide, 99.995% (metals basis)
Description
For vacuum deposition. In the production of catalysts. Used as part of the manufacturing of semiconductor devices. Has been shown to demonstrate catalytic properties and has also been used as an insulating barrier in tight junctions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1900°C |
| Physical Form | Sintered Lump |
| Quantity | 25 g |
| Odor | Odorless |
| Refractive Index | 1.92 |
| Density | 6.44 g/mL |
| Merck Index | 14,4346 |
| Solubility Information | Insoluble in water. Soluble in most acids. |
| Formula Weight | 187.44 |
| Percent Purity | 99.995% |
| Show More |
Safety and Handling
| EINECSNumber | 234-691-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only