missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) oxide, 99.99+%, (trace metal basis)

Catalog No. AC201100050
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC201100050 5 g Plastic bottle
AC201100500 50 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC201100050 Supplier Thermo Scientific Chemicals Supplier No. 201100050

CAS: 12024-21-4 | Ga2O3 | 187.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

for ultra trace analysis

Chemical Identifiers

CAS 12024-21-4
Molecular Formula Ga2O3
Molecular Weight (g/mol) 187.44
MDL Number MFCD00011020
InChI Key AJNVQOSZGJRYEI-UHFFFAOYSA-N
IUPAC Name digallium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Ga+3].[Ga+3]

Specifications

Melting Point 1740°C
Color White
Physical Form Fine Crystalline Powder
Quantity 5 g
Assay Percent Range 99.99+%
Linear Formula Ga2O3
Fieser 04,241
Merck Index 15, 4382
Solubility Information Solubility in water: insoluble in water. Other solubilities: slightly soluble in acids,alkalies
Formula Weight 187.44
Percent Purity ≥99.99%
Grade Trace Metal Basis
Packaging Plastic bottle
Specific Gravity 5.88
Density 5.8800g/mL
Chemical Name or Material Gallium(III) oxide
Show More Show Less
EINECSNumber 234-691-7

RUO – Research Use Only