missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)

Catalog No. AA1115006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1115006 5 g
AA1115014 25 g
AA1115022 100 g
AA1115036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1115006 Supplier Thermo Scientific Chemicals Supplier No. 01115006

CAS: 69365-72-6 | GaN3O9 | 255.74 g/mol

Gallium(III) nitrate hydrate is the potential precursor for the synthesis of other gallium compounds. It finds application in the preparation of gallium nitride, which is used as a solar cell array for satellites. In the field of pharmaceutical industry, it is used to treat symptomatic hypercalcemia by preventing the breakdown of bone by slowing osteoclast activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69365-72-6
Molecular Formula GaN3O9
Molecular Weight (g/mol) 255.74
MDL Number MFCD00149723
InChI Key CHPZKNULDCNCBW-UHFFFAOYSA-N
Synonym gallium iii nitrate hydrate, nitric acid, gallium salt, hydrate, ga.3no3.h2o, gallium nitrate nanoparticles, gallium-iii-nitrate hydrate, gallium iii nitrate hydrate, puratronic, gallium iii nitrate hydrate, puratronic metals basis, gallium iii nitrate hydrate, crystalline trace metals basis, gallium iii nitrate hydrate, crystals and lumps trace metals basis
PubChem CID 11550823
SMILES [Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 110°C (decomposition)
Color White
Physical Form Crystalline Aggregates
Quantity 5 g
UN Number UN1477
Merck Index 14,4346
Solubility Information Soluble in water.
IUPAC Name gallium;trinitrate;hydrate
Formula Weight 255.73 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Gallium(III) nitrate hydrate
Show More Show Less
Hazard Category H272-H315-H319
Hazard Statement GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: II
EINECSNumber 236-815-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only