missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) nitrate hydrate, 99.9% (metals basis)

Catalog No. p-3208412
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3211606 5 g
AA3211614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3211606 Supplier Thermo Scientific Chemicals Supplier No. 03211606

CAS: 69365-72-6 | GaN3O9 | 255.74 g/mol

Gallium(III) nitrate hydrate is used in the preparation of gallium nitride and other gallium compounds. It acts as a drug used to treat symptomatic hypercalcemia secondary to cancer. It finds application as an antineoplastic agent and also as an acting agent for the treatment of bone-resorptive diseases. It is involved to improve the cultural selectivity of medically significant fungi by suppression of contaminating bacterial flora.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69365-72-6
Molecular Formula GaN3O9
Molecular Weight (g/mol) 255.74
MDL Number MFCD00149723
InChI Key CHPZKNULDCNCBW-UHFFFAOYSA-N
Synonym gallium iii nitrate hydrate, nitric acid, gallium salt, hydrate, ga.3no3.h2o, gallium nitrate nanoparticles, gallium-iii-nitrate hydrate, gallium iii nitrate hydrate, puratronic, gallium iii nitrate hydrate, puratronic metals basis, gallium iii nitrate hydrate, crystalline trace metals basis, gallium iii nitrate hydrate, crystals and lumps trace metals basis
PubChem CID 11550823
SMILES [Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 110°C (decomposition)
Color White
Physical Form Lump
Quantity 5 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information Soluble in water.
IUPAC Name gallium(3+) trinitrate
Formula Weight 255.73 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance White powder/flake/crystalline/beads/lump, etc.
Chemical Name or Material Gallium(III) nitrate hydrate
Show More Show Less
Hazard Category H272-H315-H319
Precautionary Statement P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
DOTInformation DOT Class: 5.1; Packaging Group: II
EINECSNumber 236-815-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only