missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) isopropoxide, mixture of oligomers, 99%

Catalog No. AA3658303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3658303 1 g
AA3658306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3658303 Supplier Thermo Scientific Chemicals Supplier No. 03658303

CAS: 4452-61-3 | C3H8GaO | 129.819 g/mol

Gallium(III) isopropoxide can be used to accelerate the drying of paints or printing inks. It is also used as effective catalysts in the conversion of an aldehyde to an ester, ketones to secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4452-61-3
Molecular Formula C3H8GaO
Molecular Weight (g/mol) 129.819
MDL Number MFCD00144389
InChI Key JTPVILBGRKEEOP-UHFFFAOYSA-N
Synonym 2-propanol, gallium salt
PubChem CID 122197675
IUPAC Name gallium;propan-2-ol
SMILES CC(C)O.[Ga]

Specifications

Melting Point 72°C
Color Yellow
Density 1.35 g/mL
Boiling Point 120°C (1 mm)
Flash Point 68°C (154°F)
Linear Formula Ga(OC3H7)3
Quantity 1 g
UN Number UN1993
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 246.98
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Gallium(III) isopropoxide, Mixture of oligomers
Show More Show Less
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H226-H315-H319
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only