missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) ethoxide

CAS: 2572-25-0 | C6H18GaO3 | 207.93 g/mol

$192.42 - $692.00

Chemical Identifiers

CAS 2572-25-0
Molecular Formula C6H18GaO3
Molecular Weight (g/mol) 207.93
MDL Number MFCD01866730
InChI Key JWISYUBLQXUTOS-UHFFFAOYSA-N
Synonym gallium iii ethoxide
PubChem CID 131876769
IUPAC Name ethanol;gallium
SMILES [Ga].CCO.CCO.CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4485903
View Documents
Thermo Scientific Chemicals
04485903
1 g
Each for $192.42
Only null left
 
AA4485906
View Documents
Thermo Scientific Chemicals
04485906
5 g
Each for $692.00
Only null left
 
Description

Description

Gallium(III) ethoxide cohydrolysis with Te alkoxides on PMMA yields heat-mode erasable optical memory. It is also used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2572-25-0
207.93
JWISYUBLQXUTOS-UHFFFAOYSA-N
131876769
[Ga].CCO.CCO.CCO
C6H18GaO3
MFCD01866730
gallium iii ethoxide
ethanol;gallium

Specifications

2572-25-0
1.25 g/mL
C6H18GaO3
MFCD01866730
Moisture sensitive
Soluble in toluene.
[Ga].CCO.CCO.CCO
207.93
204.9
Gallium (III) ethoxide
Yellow
68°C (154°F)
Ga(OC2H5)3·xHOCH2CH3
1 g
gallium iii ethoxide
JWISYUBLQXUTOS-UHFFFAOYSA-N
ethanol;gallium
131876769
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c

H227-H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only