missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) 2,4-pentanedionate, 99.99% (metals basis)

Catalog No. p-7051836
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4208606 5 g
AA4208614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4208606 Supplier Thermo Scientific Chemicals Supplier No. 04208606
Only null left

CAS: 14405-43-7 | C15H21GaO6 | 367.05 g/mol

It is used in research on Ga-containing materials. Gallium oxide thin films can be created with atomic layer epitaxy (ALE) by combining gallium acetylacetonate with either water or ozone as the precursor. It can also be used for the synthesis of high purity gallium nitride nano-wires and nano-needles at low temperatures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14405-43-7
Molecular Formula C15H21GaO6
Molecular Weight (g/mol) 367.05
MDL Number MFCD00013492
InChI Key IMMRYVXLCAMRNK-UHFFFAOYSA-N
Synonym gallium iii acetylacetonate, gallium iii 2,4-pentanedionate
PubChem CID 71311502
IUPAC Name (Z)-4-[[(Z)-4-oxopent-2-en-2-yl]oxy-[(E)-4-oxopent-2-en-2-yl]oxygallanyl]oxypent-3-en-2-one
SMILES [Ga+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Melting Point 196°C to 198°C (decomposition)
Physical Form Powder
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 367.05
Percent Purity 99.99%
Odor Odorless
Density 1.42 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) 2,4-pentanedionate
Hazard Category H302+H312+H332-H315-H319-H335-H351
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
EINECSNumber 238-377-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only