missing translation for 'onlineSavingsMsg'
Learn More

Gallium arsenide, 99.999% (metals basis)

Catalog No. AA8845818
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8845818 50 g
AA8845804 2 g
AA8845809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA8845818 Supplier Thermo Scientific Chemicals Supplier No. 08845818

For Semiconductors (Transistors, Lasers, Solar Cells) | CAS: 1303-00-0 | AsGa | 144.645 g/mol

Semiconductors (transistors, lasers, solar cells)Gallium arsenide is used in semiconductor applications. It is also used in the manufacture of devices such as microwave frequency integrated circuits, Gunn diodes, monolithic microwave integrated circuits, infrared light-emitting diodes, laser diodes and optical windows. Further, it is used for single-crystalline thin film solar cells and multi-junction solar cells. In addition, it is used for the detection of X-rays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1303-00-0
Molecular Formula AsGa
Molecular Weight (g/mol) 144.645
MDL Number MFCD00011017
InChI Key JBRZTFJDHDCESZ-UHFFFAOYSA-N
Synonym gallium arsenide, gallium arsenide gaas, gallium monoarsenide, gallium monoarsenide gaas, unii-27fc46ga44, ccris 4020, arsinogallane, gallium arsenide trace metals basis 1g, gallium arsenide, pieces trace metals basis, gallium arsenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick
PubChem CID 14770
IUPAC Name gallanylidynearsane
SMILES [Ga]#[As]

Specifications

Melting Point 1238°C
Color Gray
Physical Form Polycrystalline Pieces
Quantity 50 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Soluble in hydrochloric acid. Insoluble in water,ethanol,methanol and acetone.
Formula Weight 144.64
Odor Odorless
Density 5.31
Chemical Name or Material Gallium arsenide
Hazard Category H301+H331-H350
Hazard Statement When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
DOTInformation DOT Class: 6.1, Packing Group: II
EINECSNumber 215-114-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only